CAS RN 18531-99-2
(S)-(-)-1,1'-Bi(2-naphthol), 99%
CAS: 18531-99-2 Molecular Formula: C20H14O2 Molecular Weight (g/mol): 286.33 MDL Number: MFCD00004068 InChI Key: PPTXVXKCQZKFBN-UHFFFAOYSA-N Synonym: 1,1'-bi-2-naphthol,s---1,1'-bi-2-naphthol,r-+-1,1'-bi-2-naphthol,1,1'-binaphthalene-2,2'-diol,binol,1,1'-binaphthyl-2,2'-diol,2,2'-dinaphthol,beta-binaphthol,r-1,1'-bi-2-naphthol,s-1,1'-binaphthalene-2,2'-diol PubChem CID: 11762 IUPAC Name: 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol SMILES: C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
(S)-(-)-1,1'-Bi-2-naphthol, 99%, Thermo Scientific Chemicals
CAS: 18531-99-2 InChI Key: PPTXVXKCQZKFBN-UHFFFAOYSA-N Synonym: 1,1'-bi-2-naphthol,s---1,1'-bi-2-naphthol,r-+-1,1'-bi-2-naphthol,1,1'-binaphthalene-2,2'-diol,binol,1,1'-binaphthyl-2,2'-diol,2,2'-dinaphthol,beta-binaphthol,r-1,1'-bi-2-naphthol,s-1,1'-binaphthalene-2,2'-diol PubChem CID: 11762 IUPAC Name: 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol SMILES: C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
(S)-(-)-1,1'-Bi-2-naphthol 98.0+%, TCI America™
CAS: 18531-99-2 Molecular Formula: C20H14O2 Molecular Weight (g/mol): 286.33 MDL Number: MFCD00004068 InChI Key: PPTXVXKCQZKFBN-UHFFFAOYSA-N Synonym: 1,1'-bi-2-naphthol,s---1,1'-bi-2-naphthol,r-+-1,1'-bi-2-naphthol,1,1'-binaphthalene-2,2'-diol,binol,1,1'-binaphthyl-2,2'-diol,2,2'-dinaphthol,beta-binaphthol,r-1,1'-bi-2-naphthol,s-1,1'-binaphthalene-2,2'-diol PubChem CID: 11762 IUPAC Name: 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol SMILES: C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O
Sigma Aldrich (S)-(-)-1,1'-Bi(2-Naphthol)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
S-(-)-2,2'-Dihydroxy-1,1'-dinaphthyl, 99%, Thermo Scientific™
CAS: 18531-99-2 Molecular Formula: C20H14O2 Molecular Weight (g/mol): 286.33 MDL Number: MFCD00004068 InChI Key: PPTXVXKCQZKFBN-UHFFFAOYSA-N Synonym: 1,1'-bi-2-naphthol,s---1,1'-bi-2-naphthol,r-+-1,1'-bi-2-naphthol,1,1'-binaphthalene-2,2'-diol,binol,1,1'-binaphthyl-2,2'-diol,2,2'-dinaphthol,beta-binaphthol,r-1,1'-bi-2-naphthol,s-1,1'-binaphthalene-2,2'-diol PubChem CID: 11762 IUPAC Name: 1-(2-hydroxynaphthalen-1-yl)naphthalen-2-ol SMILES: C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)O)O