accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18542-42-2

IUPAC Name: 2-(methylsulfanyl)ethan-1-aminium
Synonyms: 2-(methylsulfanyl)ethanaminium
Molecular Weight (g/mol): 92.18
Molecular Formula: C3H10NS
InChi Key: CYWGSFFHHMQKET-UHFFFAOYSA-O
SMILES: CSCC[NH3+]
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (6)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)