accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185626-73-7

IUPAC Name: (3-bromophenyl)triphenylsilane
Synonyms: (3-bromophenyl)triphenylsilane
Molecular Weight (g/mol): 415.41
Molecular Formula: C24H19BrSi
InChi Key: WTYKIOBQNRWDQP-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results