CAS RN 185669-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Kir2.1 Inhibitor, ML133
The Kir2.1 Inhibitor, ML133, also referenced under CAS 185669-79-8, controls the biological activity of Kir2.1. This small molecule/inhibitor is primarily used for Cancer applications.