accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 185690-41-9

IUPAC Name: N1-(naphthalen-2-yl)-N4,N4-bis({4-[(naphthalen-2-yl)(phenyl)amino]phenyl})-N1-phenylbenzene-1,4-diamine
Synonyms: N1,N1-bis({4-[naphthalen-2-yl(phenyl)amino]phenyl})-N4-(naphthalen-2-yl)-N4-phenylbenzene-1,4-diamine
Molecular Weight (g/mol): 897.14
Molecular Formula: C66H48N4
InChi Key: KDOQMLIRFUVJNT-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=C(C=C1)N(C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results