accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OH3N

CAS RN 1857-19-8

IUPAC Name: (dimethylcarbamoyl)methanaminium
Synonyms: (dimethylcarbamoyl)methanaminium
Molecular Weight (g/mol): 103.14
Molecular Formula: C4H11N2O
InChi Key: KNVRBEGQERGQRP-UHFFFAOYSA-O
SMILES: CN(C)C(=O)C[NH3+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)