accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNOOOCH3OOCH3OOCH3OOCH3OOOOOOH3COOOOOOCH3

CAS RN 1858223-85-4

IUPAC Name: methyl 4-(acetyloxy)-2-{[3,5-bis(benzoyloxy)-2-[(benzoyloxy)methyl]-6-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-acetamido-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Synonyms: methyl 4-(acetyloxy)-2-{[3,5-bis(benzoyloxy)-2-[(benzoyloxy)methyl]-6-(4-methoxyphenoxy)oxan-4-yl]oxy}-5-acetamido-6-[1,2,3-tris(acetyloxy)propyl]oxane-2-carboxylate
Molecular Weight (g/mol): 1072.04
Molecular Formula: C54H57NO22
InChi Key: DNICDVYFYXDKKN-UHFFFAOYNA-N
SMILES: COC(=O)C1(CC(OC(C)=O)C(NC(C)=O)C(O1)C(OC(C)=O)C(COC(C)=O)OC(C)=O)OC1C(OC(=O)C2=CC=CC=C2)C(COC(=O)C2=CC=CC=C2)OC(OC2=CC=C(OC)C=C2)C1OC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results