accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNOCH3Br

CAS RN 1858241-84-5

IUPAC Name: 3-(4-bromophenyl)-4-[(4-methoxyphenyl)methyl]-5-methyl-4H-1,2,4-triazole
Synonyms: 3-(4-bromophenyl)-4-[(4-methoxyphenyl)methyl]-5-methyl-1,2,4-triazole
Molecular Weight (g/mol): 358.24
Molecular Formula: C17H16BrN3O
InChi Key: YMJHVVWEYLBWNG-UHFFFAOYSA-N
SMILES: COC1=CC=C(CN2C(C)=NN=C2C2=CC=C(Br)C=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results