accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSNNOO

CAS RN 1858241-96-9

IUPAC Name: (9H-fluoren-9-yl)methyl 3-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]-1H-indole-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 3-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]indole-1-carboxylate
Molecular Weight (g/mol): 494.57
Molecular Formula: C29H22N2O4S
InChi Key: ZQNWSUMADTWVPW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CSC(=N1)C1=CN(C(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results