accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1858251-56-5

NNONOO

CAS RN 1858251-56-5

IUPAC Name: (9H-fluoren-9-yl)methyl 3-(5-cyclopropyl-1,2,4-oxadiazol-3-yl)-1H-indole-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 3-(5-cyclopropyl-1,2,4-oxadiazol-3-yl)indole-1-carboxylate
Molecular Weight (g/mol): 447.49
Molecular Formula: C28H21N3O3
InChi Key: PFVLQGHFBHHTCJ-UHFFFAOYSA-N
SMILES: O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)N1C=C(C2=NOC(=N2)C2CC2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results