accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNSN

CAS RN 1858251-64-5

IUPAC Name: 2-(3-methylphenyl)-4-phenyl-1,3-thiazole-5-carbonitrile
Synonyms: 2-(3-methylphenyl)-4-phenyl-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol): 276.36
Molecular Formula: C17H12N2S
InChi Key: DUWIGOHYDXAKEV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C1=NC(=C(S1)C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results