accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1858252-00-2

IUPAC Name: 6-benzoyl-3-(thiophen-2-yl)-1,2-dihydroquinoxalin-2-one
Synonyms: 6-benzoyl-3-(thiophen-2-yl)-1H-quinoxalin-2-one
Molecular Weight (g/mol): 332.38
Molecular Formula: C19H12N2O2S
InChi Key: DAONFPVIAXDGSI-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C1=CC=C2NC(=O)C(=NC2=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results