accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3SOONONNHOONOO

CAS RN 1858255-08-9

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 6-{[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl]amino}hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 6-{[7-(dimethylsulfamoyl)-2,1,3-benzoxadiazol-4-yl]amino}hexanoate
Molecular Weight (g/mol): 453.47
Molecular Formula: C18H23N5O7S
InChi Key: MMPAJYBUTABRFE-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(NCCCCCC(=O)ON2C(=O)CCC2=O)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results