accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONNONCl

CAS RN 1858255-72-7

IUPAC Name: (9H-fluoren-9-yl)methyl 3-[5-(chloromethyl)-1,2,4-oxadiazol-3-yl]-1H-indole-1-carboxylate
Synonyms: 9H-fluoren-9-ylmethyl 3-[5-(chloromethyl)-1,2,4-oxadiazol-3-yl]indole-1-carboxylate
Molecular Weight (g/mol): 455.9
Molecular Formula: C26H18ClN3O3
InChi Key: WRXXFUJGWLIKEU-UHFFFAOYSA-N
SMILES: ClCC1=NC(=NO1)C1=CN(C(=O)OCC2C3=CC=CC=C3C3=CC=CC=C23)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results