accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNNOHNH2(E/Z)

CAS RN 1858264-55-7

IUPAC Name: N'-hydroxy-4-phenyl-2-(pyridin-3-yl)-1,3-thiazole-5-carboximidamide
Synonyms: N'-hydroxy-4-phenyl-2-(pyridin-3-yl)-1,3-thiazole-5-carboximidamide
Molecular Weight (g/mol): 296.35
Molecular Formula: C15H12N4OS
InChi Key: XYSMWYBSBQJLBN-UHFFFAOYSA-N
SMILES: NC(=NO)C1=C(N=C(S1)C1=CN=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results