accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18583-89-6

IUPAC Name: methyl 3-amino-2-methylbenzoate
Synonyms: methyl 3-amino-2-methylbenzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: ZOOQFAUFPWXUMI-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)C(N)=CC=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)