CAS RN 185855-30-5
IUPAC Name:
1,2,4-trimethyl cyclohexane-1,2,4-tricarboxylate
Synonyms:
1,2,4-trimethyl cyclohexane-1,2,4-tricarboxylate
Molecular Weight (g/mol):
258.27
Molecular Formula:
C12H18O6
InChi Key:
DEIJHCATUHBNIH-UHFFFAOYNA-N
SMILES:
COC(=O)C1CCC(C(C1)C(=O)OC)C(=O)OC
1
–
1
of
1
results
1
Trimethyl 1,2,4-Cyclohexanetricarboxylate (cis- and trans- mixture) 97.0+%, TCI America™
CAS: 185855-30-5 Molecular Formula: C12H18O6 Molecular Weight (g/mol): 258.27 InChI Key: DEIJHCATUHBNIH-UHFFFAOYSA-N Synonym: 1,2,4-Cyclohexanetricarboxylic Acid Trimethyl Ester PubChem CID: 12616206 IUPAC Name: trimethyl cyclohexane-1,2,4-tricarboxylate SMILES: COC(=O)C1CCC(C(C1)C(=O)OC)C(=O)OC