accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 18594-05-3

IUPAC Name: 1-(4-cyclohexylphenyl)ethan-1-one
Synonyms: 1-(4-cyclohexylphenyl)ethanone
Molecular Weight (g/mol): 202.3
Molecular Formula: C14H18O
InChi Key: MSDQNIRGPBARGC-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)C1CCCCC1
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)