accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONH2H3C

CAS RN 18595-14-7

IUPAC Name: methyl 4-amino-3-methylbenzoate
Synonyms: methyl 4-amino-3-methylbenzoate
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: ZHIPSMIKSRYZFV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(N)C(C)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)