accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18598-74-8

IUPAC Name: methyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Synonyms: methyl L-isoleucinate hydrochloride
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: GGTBEWGOPAFTTH-GEMLJDPKSA-N
SMILES: Cl.CC[C@H](C)[C@H](N)C(=O)OC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)