accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClClF

CAS RN 185985-40-4

IUPAC Name: 2,3-dichloro-5-fluoropyridine
Synonyms: 2,3-dichloro-5-fluoropyridine
Molecular Weight (g/mol): 165.98
Molecular Formula: C5H2Cl2FN
InChi Key: YIYIQENCPIISLL-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=C(Cl)N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results