accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CSiH3CCH3

CAS RN 185990-03-8

IUPAC Name: dimethyl(phenyl)(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane
Synonyms: dimethyl(phenyl)(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)silane
Molecular Weight (g/mol): 262.23
Molecular Formula: C14H23BO2Si
InChi Key: ARMSAQNLTKGMGM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)[Si](C)(C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)