accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1860-26-0

IUPAC Name: tris(2-ethylhexyl)amine
Synonyms: tris(2-ethylhexyl)amine
Molecular Weight (g/mol): 353.68
Molecular Formula: C24H51N
InChi Key: BZUDVELGTZDOIG-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN(CC(CC)CCCC)CC(CC)CCCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)