accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18600-42-5

ClH3NNOO

CAS RN 18600-42-5

IUPAC Name: (4-nitrophenyl)methanaminium chloride
Synonyms: (4-nitrophenyl)methanaminium chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: SMIXZZMSWYOQPW-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]CC1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (5)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)