accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NFNOO

CAS RN 18605-16-8

IUPAC Name: 6-fluoro-2-methyl-3-nitropyridine
Synonyms: 6-fluoro-2-methyl-3-nitropyridine
Molecular Weight (g/mol): 156.12
Molecular Formula: C6H5FN2O2
InChi Key: JVLRFRZVCOWBDY-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(F)=N1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)