accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18618-55-8

IUPAC Name: trichlorocerium heptahydrate
Synonyms: cerium chloride heptahydrate
Molecular Weight (g/mol): 372.57
Molecular Formula: CeCl3H14O7
InChi Key: KPZSTOVTJYRDIO-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.O.Cl[Ce](Cl)Cl
Quantity 
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (6)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (8)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (6)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (2)
  • (8)
  • (2)
  • (3)