accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186259-63-2

IUPAC Name: 2,7-dibromo-9,9-diphenyl-9H-fluorene
Synonyms: 2,7-dibromo-9,9-diphenylfluorene
Molecular Weight (g/mol): 476.21
Molecular Formula: C25H16Br2
InChi Key: AJYDOCCGNIBJBY-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C2(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results