CAS RN 18628-07-4
IUPAC Name:
9H-3,9'-bicarbazole
Synonyms:
9H-3,9'-bicarbazole
Molecular Weight (g/mol):
332.41
Molecular Formula:
C24H16N2
InChi Key:
FHJJVSJWFYYPAC-UHFFFAOYSA-N
SMILES:
N1C2=C(C=CC=C2)C2=C1C=CC(=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2
1
–
1
of
1
results
1
3,9'-Bicarbazole 98.0+%, TCI America™
CAS: 18628-07-4 Molecular Formula: C24H16N2 Molecular Weight (g/mol): 332.406 MDL Number: MFCD27644690 InChI Key: FHJJVSJWFYYPAC-UHFFFAOYSA-N PubChem CID: 13236022 IUPAC Name: 3-carbazol-9-yl-9H-carbazole SMILES: C1=CC=C2C(=C1)C3=C(N2)C=CC(=C3)N4C5=CC=CC=C5C6=CC=CC=C64