accessibility menu, dialog, popup

UserName

Viewing profile as user:
HBrNH2NH2OHO

CAS RN 18635-45-5

IUPAC Name: hydrogen 2,3-diaminopropanoic acid bromide
Synonyms: hydrogen 2,3-diaminopropanoic acid bromide
Molecular Weight (g/mol): 185.02
Molecular Formula: C3H9BrN2O2
InChi Key: IZNPALADKWAZMW-UHFFFAOYNA-N
SMILES: [H+].[Br-].NCC(N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results