accessibility menu, dialog, popup

UserName

Viewing profile as user:
Br

CAS RN 18648-66-3

IUPAC Name: 1-bromo-4-(2,2-diphenylethenyl)benzene
Synonyms: 1-bromo-4-(2,2-diphenylethenyl)benzene
Molecular Weight (g/mol): 335.24
Molecular Formula: C20H15Br
InChi Key: HUCFDUOIMSRYAA-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results