accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186496-59-3

CH3FMgBr

CAS RN 186496-59-3

IUPAC Name: bromo(5-fluoro-2-methylphenyl)magnesium
Synonyms: bromo(5-fluoro-2-methylphenyl)magnesium
Molecular Weight (g/mol): 213.33
Molecular Formula: C7H6BrFMg
InChi Key: XKMSJPVAJJFYFU-UHFFFAOYSA-M
SMILES: CC1=CC=C(F)C=C1[Mg]Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results