accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186498-02-2

IUPAC Name: [4-(morpholin-4-yl)phenyl]boronic acid
Synonyms: 4-(morpholin-4-yl)phenylboronic acid
Molecular Weight (g/mol): 207.04
Molecular Formula: C10H14BNO3
InChi Key: WHDIUBHAKZDSJL-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results