accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186550-26-5

IUPAC Name: 3'-methoxy-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms: 3'-methoxy-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol): 282.74
Molecular Formula: C13H11ClO3S
InChi Key: DVQBUJTYQXKLKZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C1=CC=C(C=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results