accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18657-57-3

IUPAC Name: 3-[2-(1H-inden-3-yl)ethyl]-1H-indene
Synonyms: 1-[2-(3H-inden-1-yl)ethyl]-3H-indene
Molecular Weight (g/mol): 258.36
Molecular Formula: C20H18
InChi Key: CQAQBIQKEFJNRZ-UHFFFAOYSA-N
SMILES: C(CC1=CCC2=CC=CC=C12)C1=CCC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results