accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186590-26-1

IUPAC Name: 5-bromo-2,3-difluorophenol
Synonyms: 5-bromo-2,3-difluorophenol
Molecular Weight (g/mol): 208.99
Molecular Formula: C6H3BrF2O
InChi Key: QAHCQGXGAYRHHW-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=CC(F)=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)