accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186593-48-6

CH3NFNH2

CAS RN 186593-48-6

IUPAC Name: 6-fluoro-5-methylpyridin-3-amine
Synonyms: 6-fluoro-5-methylpyridin-3-amine
Molecular Weight (g/mol): 126.13
Molecular Formula: C6H7FN2
InChi Key: KADQKKYTJHXBSL-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CN=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results