accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCH3CH3H3CCH3SOOO

CAS RN 1866-13-3

IUPAC Name: trimethyl[2-(propanoyloxy)ethyl]azanium 4-methylbenzene-1-sulfonate
Synonyms: β-methylacetylcholine tosylate
Molecular Weight (g/mol): 331.43
Molecular Formula: C15H25NO5S
InChi Key: QEWHHEJZLZTUQT-UHFFFAOYSA-M
SMILES: CCC(=O)OCC[N+](C)(C)C.CC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results