accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOHOOOOH3COOCH3OOCH3

CAS RN 186605-76-5

IUPAC Name: 1,3-dimethyl 5-[(3-{[3,5-bis(methoxycarbonyl)phenoxy]methyl}-5-hydroxyphenyl)methoxy]benzene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 5-({3-[3,5-bis(methoxycarbonyl)phenoxymethyl]-5-hydroxyphenyl}methoxy)benzene-1,3-dicarboxylate
Molecular Weight (g/mol): 538.51
Molecular Formula: C28H26O11
InChi Key: USUINPRIDFWVMZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC(OCC2=CC(COC3=CC(=CC(=C3)C(=O)OC)C(=O)OC)=CC(O)=C2)=C1)C(=O)OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results