accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2ONaNaNaOONOOOO

CAS RN 18662-53-8

IUPAC Name: trisodium 2-[bis(carboxylatomethyl)amino]acetate hydrate
Synonyms: trisodium hydrate nitrilotriacetate
Molecular Weight (g/mol): 275.1
Molecular Formula: C6H8NNa3O7
InChi Key: HERBOKBJKVUALN-UHFFFAOYSA-K
SMILES: O.[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)