CAS RN 18666-26-7
CAS RN 18666-26-7
IUPAC Name:
bis(1H-inden-1-yl)dimethylsilane
Synonyms:
bis(1H-inden-1-yl)dimethylsilane
Molecular Weight (g/mol):
288.47
Molecular Formula:
C20H20Si
InChi Key:
DXYTUIWIDDBVLU-UHFFFAOYNA-N
SMILES:
C[Si](C)(C1C=CC2=CC=CC=C12)C1C=CC2=CC=CC=C12
1
–
1
of
1
results
1
Thermo Scientific Chemicals Bis(1H-inden-1-yl)dimethylsilane, 97%, Thermo Scientific™
CAS: 18666-26-7 MDL Number: MFCD00674015 InChI Key: DXYTUIWIDDBVLU-UHFFFAOYSA-N Synonym: bis indenyl dimethylsilane,di 1h-inden-1-yl dimethylsilane,bis 1h-inden-1-yl-dimethylsilane,1h-indene,1,1'-dimethylsilylene bis,bis 1h-inden-1-yl dimethylsilane,dimethylbisindenylsilane,acmc-1bvug,dimethyldi 1-indenyl silane,dimethylbis 1-indenyl silane,dimethylbis indenyl silane PubChem CID: 599429 IUPAC Name: bis(1H-inden-1-yl)-dimethylsilane SMILES: C[Si](C)(C1C=CC2=CC=CC=C12)C3C=CC4=CC=CC=C34