accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18666-68-7

IUPAC Name: ethenyltriphenylsilane
Synonyms: ethenyltriphenylsilane
Molecular Weight (g/mol): 286.45
Molecular Formula: C20H18Si
InChi Key: OVOIHGSHJGMSMZ-UHFFFAOYSA-N
SMILES: C=C[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results