accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186704-03-0

IUPAC Name: 1-tert-butyl 2-methyl 2,3-dihydro-1H-indole-1,2-dicarboxylate
Synonyms: 1-tert-butyl 2-methyl 2,3-dihydroindole-1,2-dicarboxylate
Molecular Weight (g/mol): 277.32
Molecular Formula: C15H19NO4
InChi Key: JWTYDRKOSWALHD-UHFFFAOYNA-N
SMILES: COC(=O)C1CC2=CC=CC=C2N1C(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results