accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOH

CAS RN 186788-38-5

IUPAC Name: 10-(1H-imidazol-1-yl)decan-1-ol
Synonyms: 10-(imidazol-1-yl)decan-1-ol
Molecular Weight (g/mol): 224.35
Molecular Formula: C13H24N2O
InChi Key: XFIJCEYDSGZRHB-UHFFFAOYSA-N
SMILES: OCCCCCCCCCCN1C=CN=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results