accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClO

CAS RN 1871-76-7

IUPAC Name: 2,2-diphenylacetyl chloride
Synonyms: diphenyl-acetyl chloride
Molecular Weight (g/mol): 230.69
Molecular Formula: C14H11ClO
InChi Key: MSYLETHDEIJMAF-UHFFFAOYSA-N
SMILES: ClC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)