accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1871-96-1

IUPAC Name: decanedinitrile
Synonyms: decanedinitrile
Molecular Weight (g/mol): 164.25
Molecular Formula: C10H16N2
InChi Key: DFJYZCUIKPGCSG-UHFFFAOYSA-N
SMILES: N#CCCCCCCCCC#N
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)