CAS RN 18720-35-9
IUPAC Name:
4-(methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid
Synonyms:
4-(methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid
Molecular Weight (g/mol):
212.25
Molecular Formula:
C11H16O4
InChi Key:
KBZVAQVEHVGIEI-UHFFFAOYSA-N
SMILES:
COC(=O)C12CCC(CC1)(CC2)C(O)=O
1
–
2
of
2
results
2
Sigma Aldrich 4-(Methoxycarbonyl)bicyclo[2.2.2]octane-1-carboxylic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.