accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOOOONHNNHNH(S)H3CCH3H3CCH3CH3H3C

CAS RN 187283-25-6

IUPAC Name: (2S)-3-(4-{[bis({[(tert-butoxy)carbonyl]amino})methylidene]amino}phenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Synonyms: (2S)-3-[4-({bis[(tert-butoxycarbonyl)amino]methylidene}amino)phenyl]-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 644.73
Molecular Formula: C35H40N4O8
InChi Key: CIEGZABLYKOZLG-NDEPHWFRSA-N
SMILES: CC(C)(C)OC(=O)NC(NC(=O)OC(C)(C)C)=NC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)