accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1873-77-4

IUPAC Name: 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
Synonyms: 1,1,1,3,3,3-hexamethyl-2-(trimethylsilyl)trisilane
Molecular Weight (g/mol): 248.66
Molecular Formula: C9H28Si4
InChi Key: SQMFULTZZQBFBM-UHFFFAOYSA-N
SMILES: C[Si](C)(C)[SiH]([Si](C)(C)C)[Si](C)(C)C
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)