accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1873-89-8

IUPAC Name: 2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Synonyms: 2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptan-4-yl
Molecular Weight (g/mol): 295.65
Molecular Formula: C9H27O3Si4
InChi Key: XAASNKQYFKTYTR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](O[Si](C)(C)C)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results