accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrSiBr

CAS RN 18733-91-0

IUPAC Name: bis(4-bromophenyl)diphenylsilane
Synonyms: bis(4-bromophenyl)diphenylsilane
Molecular Weight (g/mol): 494.3
Molecular Formula: C24H18Br2Si
InChi Key: UTEBJTKMYMQRIF-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results